Dimethyl-trans-cyclohexan-1,4-dicarboxylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dimethyl trans-1,4-cyclohexanedicarboxylate, Dimethyl trans-1,4-Cyclohexanedicarboxylate

M-Clarity™ Qualitätsstufe = MQ100

366684-1G
SIAL366684-1GEA 79 EUR
SIAL366684-1G
Dimethyl-trans-cyclohexan-1,4-dicarboxylat, Sigma-Aldrich®
Dimethyl-trans-cyclohexan-1,4-dicarboxylat
Formel: C₁₀H₁₆O₄
Molecular Weight: 200,23 g/mol
MDL: MFCD00063917
CAS-Nummer: 3399-22-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR