Methyl-2-methyl-3-nitrobenzoat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Methyl 2-methyl-3-nitrobenzoate
M-Clarity™ Qualitätsstufe = MQ100
366765-25G
SIAL366765-25GEA
50.5
EUR
SIAL366765-25G
Methyl-2-methyl-3-nitrobenzoat, Sigma-Aldrich®
Methyl-2-methyl-3-nitrobenzoat
Formel:
C₉H₉NO₄ Molecular Weight: 195,17 g/mol |
MDL:
MFCD00082564 CAS-Nummer: 59382-59-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...