Zirkonium(IV)sulfat Hydrat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
366773-25G
SIAL366773-25GEA
430
EUR
SIAL366773-25G
Zirkonium(IV)sulfat Hydrat, Sigma-Aldrich®
Zirkonium(IV)sulfat Hydrat
Formel:
O₈S₂Zr·H₂O |
MDL:
MFCD00167043 CAS-Nummer: 34806-73-0 EINECS: 238-694-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...