N-Ethylpyridin-4-methylamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-(4-Pyridylmethyl)ethylamine, 4-(Ethylaminomethyl)pyridine

M-Clarity™ Qualitätsstufe = MQ100

367095-10G
SIAL367095-10GEA 86.9 EUR
SIAL367095-10G
N-Ethylpyridin-4-methylamin, Sigma-Aldrich®
N-Ethylpyridin-4-methylamin
Formel: C₈H₁₂N₂
Molecular Weight: 136,20 g/mol
MDL: MFCD00023632
CAS-Nummer: 33403-97-3
EINECS: 251-506-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR