(Tributylstannyl)trimethylsilan, Sigma-Aldrich®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ200

367265-5G
SIAL367265-5GEA 245 EUR
SIAL367265-5G
(Tributylstannyl)trimethylsilan, Sigma-Aldrich®
(Tributylstannyl)trimethylsilan
Formel: C₁₅H₃₆SiSn
MDL: MFCD00054903
CAS-Nummer: 17955-46-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR