1,1,1,3,3,3-Hexafluorisopropylacrylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,1,1,3,3,3-Hexafluoropropan-2-yl acrylate

M-Clarity™ Qualitätsstufe = MQ100

367656-5G
SIAL367656-5GEA 65 EUR
SIAL367656-5G
1,1,1,3,3,3-Hexafluorisopropylacrylat, Sigma-Aldrich®
1,1,1,3,3,3-Hexafluorisopropylacrylat
Formel: C₆H₄F₆O₂
Molecular Weight: 222,09 g/mol
MDL: MFCD00040104
CAS-Nummer: 2160-89-6
EINECS: 218-479-1
UN: 3272
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR