1,1,1,3,3,3-Hexafluorisopropylmethacrylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Hexafluoroisopropyl methacrylate, 1,1,1,3,3,3-Hexafluoroisopropylmethacrylate

M-Clarity™ Qualitätsstufe = MQ100

367664-5G
SIAL367664-5GEA 92.8 EUR
SIAL367664-5G
1,1,1,3,3,3-Hexafluorisopropylmethacrylat, Sigma-Aldrich®
1,1,1,3,3,3-Hexafluorisopropylmethacrylat
Formel: C₇H₆F₆O₂
Molecular Weight: 236.12 g/mol
MDL: MFCD00040105
CAS-Nummer: 3063-94-3
EINECS: 221-309-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR