4-Methyl-2-nitroanisol, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 4-Methoxy-3-nitrotoluol, Methyl-2-nitro-p-tolylether, 1-Methoxy-4-methyl-2-nitrobenzol

367699-100ML
SIAL367699-100MLEA 140 EUR
SIAL367699-100ML
4-Methyl-2-nitroanisol, Sigma-Aldrich®
4-Methyl-2-nitroanisol
Formel: C₈H₉NO₃
Molecular Weight: 167,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00024540
CAS-Nummer: 119-10-8
EINECS: 204-296-4
UN: 2446
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR