Certriacetat Hydrat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Essigsäure Cer(III)salz Hydrat, Cer(III)acetat Hydrat
M-Clarity™ Qualitätsstufe = MQ100
367753-250G
367753-50G
SIAL367753-250GEA
258
EUR
SIAL367753-250G
SIAL367753-50G
Certriacetat Hydrat, Sigma-Aldrich®
Certriacetat Hydrat
Formel:
C₆H₉CeO₆·H₂O |
MDL:
MFCD00150533 CAS-Nummer: 206996-60-3 EINECS: 208-654-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...