8-Chinolincarbonsäure, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Quinoline-8-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

367796-250MG
SIAL367796-250MGEA 65.1 EUR
SIAL367796-250MG
8-Chinolincarbonsäure, Sigma-Aldrich®
8-Chinolincarbonsäure
Formel: C₁₀H₇NO₂
Molecular Weight: 173,17 g/mol
MDL: MFCD00047619
CAS-Nummer: 86-59-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR