1,5-Diamino-9,10-anthrachinon, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
1,5-Diaminoanthrachinon
M-Clarity™ Qualitätsstufe = MQ100
367842-25G
SIAL367842-25GEA
90.3
EUR
SIAL367842-25G
1,5-Diamino-9,10-anthrachinon, Sigma-Aldrich®
1,5-Diamino-9,10-anthrachinon
Formel:
C₁₄H₁₀N₂O₂ Molecular Weight: 238,25 g/mol Schmelzpunkt: 314 °C |
MDL:
MFCD00001226 CAS-Nummer: 129-44-2 EINECS: 204-947-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...