3,5-Bis(benzyloxy)benzaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,5-Dibenzyloxybenzaldehyde

M-Clarity™ Qualitätsstufe = MQ100

368105-1G
SIAL368105-1GEA 37.6 EUR
SIAL368105-1G
3,5-Bis(benzyloxy)benzaldehyd, Sigma-Aldrich®
3,5-Bis(benzyloxy)benzaldehyd
Formel: C₂₁H₁₈O₃
Molecular Weight: 318,3658 g/mol
MDL: MFCD00075777
CAS-Nummer: 14615-72-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR