3,5-Dihydroxybenzaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

368113-1G
SIAL368113-1GEA 83.4 EUR
SIAL368113-1G
3,5-Dihydroxybenzaldehyd, Sigma-Aldrich®
3,5-Dihydroxybenzaldehyde
Formel: C₇H₆O₃
Molecular Weight: 138,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00016611
CAS-Nummer: 26153-38-8
EINECS: 247-479-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR