4,4'-(Hexafluorisopropyliden)dianilin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,2-Bis(4-aminophenyl)hexafluoropropane

368148-1G
SIAL368148-1GEA 66.4 EUR
SIAL368148-1G
4,4'-(Hexafluorisopropyliden)dianilin, Sigma-Aldrich®
4,4'-(Hexafluorisopropyliden)dianilin
Formel: C₁₅H₁₂F₆N₂
Molecular Weight: 334,2600 g/mol
MDL: MFCD00039146
CAS-Nummer: 1095-78-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR