1,3-Dichlorisochinolin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

368830-5G
SIAL368830-5GEA 154 EUR
SIAL368830-5G
1,3-Dichlorisochinolin, Sigma-Aldrich®
1,3-Dichloroisoquinoline
Formel: C₉H₅Cl₂N
Molecular Weight: 198,05 g/mol
MDL: MFCD00034750
CAS-Nummer: 7742-73-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR