3-Isochinolinol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Hydroxyisoquinoline

M-Clarity™ Qualitätsstufe = MQ100

368954-1G
SIAL368954-1GEA 32.1 EUR
SIAL368954-1G
3-Isochinolinol, Sigma-Aldrich®
3-Isochinolinol
Formel: C₉H₇NO
Molecular Weight: 145,16 g/mol
MDL: MFCD00075524
CAS-Nummer: 7651-81-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR