(tert-Butoxycarbonylmethyl)triphenylphosphoniumbromid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2-(tert-Butoxy)-2-oxoethyl)triphenylphosphonium bromide

369047-50G
SIAL369047-50GEA 250 EUR
SIAL369047-50G
(tert-Butoxycarbonylmethyl)triphenylphosphoniumbromid, Sigma-Aldrich®
(tert-Butoxycarbonylmethyl)triphenylphosphoniumbromid
Formel: C₂₄H₂₆BrO₂P
Molecular Weight: 457.35 g/mol
MDL: MFCD00075527
CAS-Nummer: 59159-39-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR