2-Brom-4-chlor-6-fluoranilin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

369063-5G
SIAL369063-5GEA 31.3 EUR
SIAL369063-5G
2-Brom-4-chlor-6-fluoranilin, Sigma-Aldrich®
2-Brom-4-chlor-6-fluoranilin
Formel: C₆H₄BrClFN
Molecular Weight: 224,46 g/mol
MDL: MFCD00070746
CAS-Nummer: 195191-47-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR