2-Brom-4,5-difluorphenol, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 1-Bromo-3,4-difluoro-6-hydroxybenzene

M-Clarity™ Qualitätsstufe = MQ100

369071-5G
SIAL369071-5GEA 50.6 EUR
SIAL369071-5G
2-Brom-4,5-difluorphenol, Sigma-Aldrich®
2-Brom-4,5-difluorphenol
Formel: C₆H₃BrF₂O
MDL: MFCD00070751
CAS-Nummer: 166281-37-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR