N,N,N',N',N''-Pentamethyldiethylentriamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N1-(2-(Dimethylamino)ethyl)-N1,N2,N2-trimethylethane-1,2-diamine, 1,1,4,7,7-Pentamethyldiethylenetriamine

M-Clarity™ Qualitätsstufe = MQ200

369497-250ML 369497-1L
SIAL369497-250MLEA 203 EUR
SIAL369497-250ML SIAL369497-1L
N,N,N',N',N''-Pentamethyldiethylentriamin, Sigma-Aldrich®
N,N,N',N',N''-Pentamethyldiethylentriamin
Formel: C₉H₂₃N₃
Molecular Weight: 173.30 g/mol
MDL: MFCD00014876
CAS-Nummer: 3030-47-5
EINECS: 221-201-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR