trans-4-(4-Pentylcyclohexyl)benzonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(trans-4-n-Pentylcyclohexyl)benzonitrile

M-Clarity™ Qualitätsstufe = MQ200

370118-1G
SIAL370118-1GEA 123 EUR
SIAL370118-1G
trans-4-(4-Pentylcyclohexyl)benzonitril, Sigma-Aldrich®
trans-4-(4-Pentylcyclohexyl)benzonitril
Formel: C₁₈H₂₅N
Molecular Weight: 255,40 g/mol
MDL: MFCD00075548
CAS-Nummer: 61204-01-1
EINECS: 262-656-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR