Methyl-3,5-dimethylbenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 3,5-dimethylbenzoate

M-Clarity™ Qualitätsstufe = MQ100

370215-5G
SIAL370215-5GEA 58.2 EUR
SIAL370215-5G
Methyl-3,5-dimethylbenzoat, Sigma-Aldrich®
Methyl-3,5-dimethylbenzoat
Formel: C₁₀H₁₂O₂
Molecular Weight: 164,21 g/mol
MDL: MFCD00077971
CAS-Nummer: 25081-39-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR