1H,1H,2H,2H-Tridecafluor-1-octanol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

370533-25G 370533-5G
SIAL370533-25GEA 164 EUR
SIAL370533-25G SIAL370533-5G
1H,1H,2H,2H-Tridecafluor-1-octanol, Sigma-Aldrich®
1H,1H,2H,2H-Tridecafluor-1-octanol
Formel: C₈H₅F₁₃O
MDL: MFCD00042143
CAS-Nummer: 647-42-7
EINECS: 211-477-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR