1H,1H,2H,2H-Tridecafluor-1-octanol, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
370533-25G
370533-5G
SIAL370533-25GEA
164
EUR
SIAL370533-25G
SIAL370533-5G
1H,1H,2H,2H-Tridecafluor-1-octanol, Sigma-Aldrich®
1H,1H,2H,2H-Tridecafluor-1-octanol
Formel:
C₈H₅F₁₃O |
MDL:
MFCD00042143 CAS-Nummer: 647-42-7 EINECS: 211-477-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...