2,2,2-Trifluor-3'-(trifluormethyl)acetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,2,2-Trifluoro-1-(3-(trifluoromethyl)phenyl)ethanone

M-Clarity™ Qualitätsstufe = MQ100

370630-5G
SIAL370630-5GEA 248 EUR
SIAL370630-5G
2,2,2-Trifluor-3'-(trifluormethyl)acetophenon, Sigma-Aldrich®
2,2,2-Trifluor-3'-(trifluormethyl)acetophenon
Formel: C₉H₄F₆O
Molecular Weight: 242,12 g/mol
MDL: MFCD00061275
CAS-Nummer: 721-37-9
UN: 1224
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR