2,2,3,4,4,4-Hexafluorbutylmethacrylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,2,3,4,4,4-Hexafluorobutyl Methacrylate

M-Clarity™ Qualitätsstufe = MQ100

371971-5G
SIAL371971-5GEA 65.7 EUR
SIAL371971-5G
2,2,3,4,4,4-Hexafluorbutylmethacrylat, Sigma-Aldrich®
2,2,3,4,4,4-Hexafluorbutylmethacrylat
Formel: C₈H₈F₆O₂
Molecular Weight: 250,14 g/mol
MDL: MFCD00042311
CAS-Nummer: 36405-47-7
UN: 3272
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR