Perfluoro(methyldecalin) Gemisch aus Isomeren ≥80%, technische Qualität

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

372439-100G
SIAL372439-100GEA 321 EUR
SIAL372439-100G
Perfluoro(methyldecalin) Gemisch aus Isomeren ≥80%, technische Qualität
Perfluoro(methyldecalin)

Perfluoro(methyldecalin) is a fluorinated solvent.


Perfluoro(methyldecalin is suitable reagent used to investigate the reactions of iodide anions and anions derived from glow discharge ionization of perfluoro(methyldecalin) with positive ions derived from poly(ethylene glycol). It is suitable reagent used for the oxidation of alcohols with the ruthenium porphyrin at higher temperatures (140ºC) and elevated oxygen pressures (50psi). Perfluoro(methyldecalin (Perfluoro-1-Methyldecalin) may be used in the preparation of undecafluoro-6-trifluoromethyltetralin via defluorination.


Contains perfluorodecalin

Formel: C₁₁F₂₀
Siedepunkt: 137…160 °C (Lit.)
Schmelzpunkt: –40 °C (Lit.)
MDL: MFCD00014326
CAS-Nummer: 51294-16-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR