Poly(2-ethyl-2-oxazolin), M.W. ~200,000, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

372854-100G
SIAL372854-100GEA 68.1 EUR
SIAL372854-100G
Poly(2-ethyl-2-oxazolin), M.W. ~200,000, Sigma-Aldrich®
Poly(2-ethyl-2-oxazolin)
Formel: (C₅H₉NO)x
Molecular Weight: 200000 g/mol
CAS-Nummer: 25805-17-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR