(S)-(+)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine, (2S)-(+)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine

M-Clarity™ Qualitätsstufe = MQ100

37289-1ML
SIAL37289-1MLEA 134 EUR
SIAL37289-1ML
(S)-(+)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazin, Sigma-Aldrich®
(S)-(+)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazin
Formel: C₉H₁₆N₂O₂
Molecular Weight: 184,2355 g/mol
MDL: MFCD00066229
CAS-Nummer: 78342-42-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR