tert-Butylchlordimethylsilan 1 mol/l in THF, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
tert-Butyl-chlordimethylsilan, Chlor-tert-butyldimethylsilan, tert-Butyldimethylchlorsilan, tert-Butyl(dimethyl)silylchlorid
M-Clarity™ Qualitätsstufe = MQ100
372951-100ML
SIAL372951-100MLEA
552
EUR
SIAL372951-100ML
tert-Butylchlordimethylsilan 1 mol/l in THF, Sigma-Aldrich®
tert-Butylchlordimethylsilan
Formel:
C₆H₁₅ClSi Molecular Weight: 150,72 g/mol |
MDL:
MFCD00000501 CAS-Nummer: 18162-48-6 EINECS: 242-042-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...