2,2-Dimethylcyclohexanon, Sigma-Aldrich®

Lieferant: Merck

373613-1G
SIAL373613-1GEA 140 EUR
SIAL373613-1G
2,2-Dimethylcyclohexanon, Sigma-Aldrich®
2,2-Dimethylcyclohexanone
Formel: C₈H₁₄O
Molecular Weight: 126,1962 g/mol
MDL: MFCD00134382
CAS-Nummer: 1193-47-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR