2,2,2-Trifluorethylmethacrylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methylpropensäure(2,2,2-trifluorethyl)ester, Methacrylsäure(2,2,2-trifluorethyl)ester

M-Clarity™ Qualitätsstufe = MQ200

373761-25G
SIAL373761-25GEA 52.4 EUR
SIAL373761-25G
2,2,2-Trifluorethylmethacrylat, Sigma-Aldrich®
2,2,2-Trifluorethylmethacrylat
Formel: C₆H₇F₃O₂
Molecular Weight: 168,12 g/mol
MDL: MFCD00013576
CAS-Nummer: 352-87-4
EINECS: 206-525-3
UN: 3272
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR