2-Isopropylimidazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Isopropyl-1H-imidazol

M-Clarity™ Qualitätsstufe = MQ100

373990-100G
SIAL373990-100GEA 67.6 EUR
SIAL373990-100G
2-Isopropylimidazol, Sigma-Aldrich®
2-Isopropylimidazol
Formel: C₆H₁₀N₂
Molecular Weight: 110,16 g/mol
MDL: MFCD00014486
CAS-Nummer: 36947-68-9
EINECS: 253-286-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR