3-Methoxypropionsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

374547-10ML
SIAL374547-10MLEA 201 EUR
SIAL374547-10ML
3-Methoxypropionsäure, Sigma-Aldrich®
3-Methoxypropionsäure
Formel: C₄H₈O₃
MDL: MFCD00042654
CAS-Nummer: 2544-06-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR