4-Brom-1H-pyrazol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

374822-10G 374822-1G
SIAL374822-10GEA 342 EUR
SIAL374822-10G SIAL374822-1G
4-Brom-1H-pyrazol, Sigma-Aldrich®
4-Bromo-1H-pyrazole
Formel: C₃H₃BrN₂
Molecular Weight: 146.97 g/mol
MDL: MFCD00075602
CAS-Nummer: 2075-45-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR