Di-tert-butyliminodicarboxylat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

375276-5G
SIAL375276-5GEA 119 EUR
SIAL375276-5G
Di-tert-butyliminodicarboxylat, Sigma-Aldrich®
Di-tert-butyliminodicarboxylat
Formel: C₁₀H₁₉NO₄
MDL: MFCD00043309
CAS-Nummer: 51779-32-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR