(1R)-(-)-Campher-10-sulfonamid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

375632-1G
SIAL375632-1GEA 59.5 EUR
SIAL375632-1G
(1R)-(-)-Campher-10-sulfonamid, Sigma-Aldrich®
(1R)-(-)-Campher-10-sulfonamid
Formel: C₁₀H₁₇NO₃S
Molecular Weight: 231,32 g/mol
MDL: MFCD00151500
CAS-Nummer: 72597-34-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR