3-Amino-2-methylbenzotrifluorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Methyl-3-(trifluoromethyl)aniline

M-Clarity™ Qualitätsstufe = MQ100

375705-1G
SIAL375705-1GEA 24.5 EUR
SIAL375705-1G
3-Amino-2-methylbenzotrifluorid, Sigma-Aldrich®
3-Amino-2-methylbenzotrifluorid
Formel: C₈H₈F₃N
Molecular Weight: 175,16 g/mol
MDL: MFCD00153216
CAS-Nummer: 54396-44-0
EINECS: 259-145-5
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR