Triisopropylchlorsilan, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Diisopropylchlorosilane
M-Clarity™ Qualitätsstufe = MQ100
375896-5G
SIAL375896-5GEA
71
EUR
SIAL375896-5G
Triisopropylchlorsilan, Sigma-Aldrich®
Chlorodiisopropylsilan
Formel:
C₆H₁₅ClSi Molecular Weight: 150,72 g/mol |
MDL:
MFCD00054896 CAS-Nummer: 2227-29-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...