Propargyltrimethylsilan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2-Propinyl)trimethylsilan, 3-(Trimethylsilyl)propin

M-Clarity™ Qualitätsstufe = MQ200

376264-1G
SIAL376264-1GEA 206 EUR
SIAL376264-1G
Propargyltrimethylsilan, Sigma-Aldrich®
Propargyltrimethylsilan
Formel: C₆H₁₂Si
Molecular Weight: 112,25 g/mol
MDL: MFCD00042922
CAS-Nummer: 13361-64-3
EINECS: 236-427-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR