(1S,3R)-(-)-Camphersäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (1S,3R)-1,2,2-Trimethylcyclopentane-1,3-dicarboxylic acid

M-Clarity™ Qualitätsstufe = MQ200

376345-1G
SIAL376345-1GEA 125 EUR
SIAL376345-1G
(1S,3R)-(-)-Camphersäure, Sigma-Aldrich®
(1S,3R)-(-)-Camphersäure
Formel: C₁₀H₁₆O₄
Molecular Weight: 200,23 g/mol
MDL: MFCD00064937
CAS-Nummer: 560-09-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR