(±)-Cbz-α-phosphonoglycintrimethylester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-Benzyloxycarbonyl-2-phosphonoglycine trimethyl ester, (±)-Benzyloxycarbonyl-α-phosphonoglycine trimethyl ester, (±)-Methyl 2-benzyloxycarbonylamino-2-(dimethoxyphosphinyl)acetate, Methyl 2-{[(benzyloxy)carbonyl]amino}-2-(dimethoxyphosphoryl)acetate, Methyl 2-(((benzyloxy)carbonyl)amino)-2-(dimethoxyphosphoryl)acetate

M-Clarity™ Qualitätsstufe = MQ200

376353-5G
SIAL376353-5GEA 139 EUR
SIAL376353-5G
(±)-Cbz-α-phosphonoglycintrimethylester, Sigma-Aldrich®
(±)-Cbz-α-phosphonoglycintrimethylester
Formel: C₁₃H₁₈NO₇P
Molecular Weight: 331,27 g/mol
MDL: MFCD00043304
CAS-Nummer: 88568-95-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR