5-Amino-1-naphthol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5,1-ANL, 5-Aminonaphthalen-1-ol

M-Clarity™ Qualitätsstufe = MQ100

376469-100G 376469-25G
SIAL376469-100GEA 297 EUR
SIAL376469-100G SIAL376469-25G
5-Amino-1-naphthol, Sigma-Aldrich®
5-Amino-1-naphthol
Formel: C₁₀H₉NO
MDL: MFCD00041826
CAS-Nummer: 83-55-6
EINECS: 201-486-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR