(1,1-Dimethoxyethyl)benzol, Sigma-Aldrich®

Lieferant: Merck

376760-5G
SIAL376760-5GEA 77.9 EUR
SIAL376760-5G
(1,1-Dimethoxyethyl)benzol, Sigma-Aldrich®
(1,1-Dimethoxyethyl)benzol
Formel: C₆H₅C(OCH₃)₂CH₃
Molecular Weight: 166,22 g/mol
MDL: MFCD00075617
CAS-Nummer: 4316-35-2
EINECS: 224-334-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2be549830e6290c28b743c9c67da48b618c0989e&vid=2be549830e6290c28b743c9c67da48b618c0989e&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR