2-Chlor-5-(trifluormethyl)benzaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

376825-5G
SIAL376825-5GEA 49.5 EUR
SIAL376825-5G
2-Chlor-5-(trifluormethyl)benzaldehyd, Sigma-Aldrich®
2-Chloro-5-(trifluoromethyl)benzaldehyde
Formel: C₈H₄ClF₃O
Molecular Weight: 208,57 g/mol
MDL: MFCD00134430
CAS-Nummer: 82386-89-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR