2-Chlor-5-(trifluormethyl)benzoesäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Chlor-α,α,α-trifluor-m-toluylsäure

M-Clarity™ Qualitätsstufe = MQ200

376833-5G
SIAL376833-5GEA 80.4 EUR
SIAL376833-5G
2-Chlor-5-(trifluormethyl)benzoesäure, Sigma-Aldrich®
2-Chlor-5-(trifluormethyl)benzoesäure
Formel: C₈H₄ClF₃O₂
Molecular Weight: 224,57 g/mol
MDL: MFCD00061094
CAS-Nummer: 657-06-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR