(R)-(+)-2-Hydroxy-3-phenylpropionsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: D-(+)-3-Phenyllactic acid, (R)-2-Hydroxy-3-phenylpropanoic acid, D(+)-Phenyllactic acid, D-​(+)​-​Phenyllactic acid

M-Clarity™ Qualitätsstufe = MQ100

376906-5G
SIAL376906-5GEA 123 EUR
SIAL376906-5G
(R)-(+)-2-Hydroxy-3-phenylpropionsäure, Sigma-Aldrich®
(R)-(+)-2-Hydroxy-3-phenylpropionsäure
Formel: C₉H₁₀O₃
MDL: MFCD00078062
CAS-Nummer: 7326-19-4
EINECS: 230-803-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR