Butylglycidether, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

377031-250ML
SIAL377031-250MLEA 65.2 EUR
SIAL377031-250ML
Butylglycidether, Sigma-Aldrich®
Butylglycidether
Formel: C₇H₁₄O₂
MDL: MFCD00005145
CAS-Nummer: 2426-08-6
EINECS: 219-376-4
UN: 3271
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR