1-Acetylindol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
N-Acetylindol, 1-Indolylmethylketon, 1-Indolyl methyl ketone, 1-(1-Indolyl)-1-ethanon, Indol-1-ylmethylketon
377104-1G
377104-5G
SIAL377104-1GEA
103
EUR
SIAL377104-1G
SIAL377104-5G
1-Acetylindol, Sigma-Aldrich®
1-Acetylindol
Formel:
C₁₀H₉NO Molecular Weight: 159.19 g/mol Siedepunkt: 97.5…101 °C (0.1 mmHg) Dichte: 1.387 g/cm³ (20 °C) |
MDL:
MFCD00038005 CAS-Nummer: 576-15-8 EINECS: 209-396-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...