Mangan(II,III)oxid, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

377473-100G
SIAL377473-100GEA 50.2 EUR
SIAL377473-100G
Mangan(II,III)oxid, Sigma-Aldrich®
Mangan(II,III)oxid
Formel: Mn₃O₄
Molecular Weight: 228,81 g/mol
MDL: MFCD00016218
CAS-Nummer: 1317-35-7
EINECS: 215-266-5
Merck Index: 13,05753

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR